But-3-yne-1-sulfonyl chloride ≥95%

Supplier: Ambeed
Danger

A1064265-1G A1064265-250MG A1064265-100MG
77570-384EA 410 USD
77570-384 77570-386 77783-052
But-3-yne-1-sulfonyl chloride ≥95%
But-3-yne-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₄H₅ClO₂S
MDL Number: MFCD19598006
CAS Number: 936644-39-2
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR