5-(Methylamino)pentan-1-ol ≥95%

Supplier: Ambeed
Danger

A1064238-1G A1064238-5G
77570-378EA 241.02 USD
77570-378 77595-726
5-(Methylamino)pentan-1-ol ≥95%
5-(Methylamino)pentan-1-ol

Precaución: for Research Use Only

Formula: C₆H₁₅NO
MW: 117.1894 g/mol
MDL Number: MFCD16696454
CAS Number: 2751-70-4
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR