2-Chloro-5-methanesulfonylpyrimidine ≥95%

Supplier: Ambeed
Warning

A1064130-50MG A1064130-100MG
77595-724EA 108.65 USD
77595-724 77619-770
2-Chloro-5-methanesulfonylpyrimidine ≥95%
2-Chloro-5-methanesulfonylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O₂S
MW: 192.6234 g/mol
MDL Number: MFCD21879442
CAS Number: 321565-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR