2-(3-Oxomorpholin-4-yl)acetic acid ≥98%

Supplier: Ambeed
Warning

A1063939-100MG A1063939-250MG
77783-042EA 48 USD
77783-042 77783-043
2-(3-Oxomorpholin-4-yl)acetic acid ≥98%
2-(3-Oxomorpholin-4-yl)acetic acid

Precaución: for Research Use Only

Formula: C₆H₉NO₄
MDL Number: MFCD16630678
CAS Number: 933692-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR