N-(5-((4-Ethylpiperazin-1-yl)methyl)pyridin-2-yl)-5-fluoro-4-(4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazol-6-yl)pyrimidin-2-amine ≥99%

Supplier: Ambeed
Warning

Synonyms: Abemaciclib

A106236-100MG A106236-1G A106236-250MG A106236-5G
77570-364EA 23.69 USD
77570-364 77610-836 77684-963 77684-964
N-(5-((4-Ethylpiperazin-1-yl)methyl)pyridin-2-yl)-5-fluoro-4-(4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazol-6-yl)pyrimidin-2-amine ≥99%
N-(5-((4-Ethylpiperazin-1-yl)methyl)pyridin-2-yl)-5-fluoro-4-(4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazol-6-yl)pyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₂₇H₃₂F₂N₈
MW: 506.59 g/mol
MDL Number: MFCD22665744
CAS Number: 1231929-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR