7-(4-(tert-Butyl)benzyl)-1,3-dimethyl-1H-purine-2,6(3H,7H)-dione ≥95%
Supplier: Ambeed
|
Warning
|
A1062272-100MG
A1062272-250MG
A1062272-50MG
77783-030EA
166
USD
77783-030
77783-031
77783-032
7-(4-(tert-Butyl)benzyl)-1,3-dimethyl-1H-purine-2,6(3H,7H)-dione ≥95%
7-(4-(tert-Butyl)benzyl)-1,3-dimethyl-1H-purine-2,6(3H,7H)-dione
Precaución: for Research Use Only
Formula:
C₁₈H₂₂N₄O₂ |
MDL Number:
MFCD02648040 CAS Number: 333415-38-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...