5-Chloropentan-2-ol ≥95%

Supplier: Ambeed
Warning

A1061457-100MG A1061457-1G
77570-358EA 66 USD
77570-358 77592-130
5-Chloropentan-2-ol ≥95%
5-Chloropentan-2-ol

Precaución: for Research Use Only

Formula: C₅H₁₁ClO
MDL Number: MFCD19231978
CAS Number: 15146-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR