2-(2-tert-Butylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%

Supplier: Ambeed
Warning

A1059875-250MG A1059875-5G
77783-008EA 83 USD
77783-008 77783-009
2-(2-tert-Butylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%
2-(2-tert-Butylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₆H₂₅BO₂
MDL Number: MFCD18734499
CAS Number: 1589507-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR