(S)-4-Hydroxypiperidin-2-one ≥95%
Supplier: Ambeed
|
Warning
|
A105949-1G
A105949-100MG
77570-340EA
249
USD
77570-340
77871-409
(S)-4-Hydroxypiperidin-2-one ≥95%
(S)-4-Hydroxypiperidin-2-one
Precaución: for Research Use Only
Formula:
C₅H₉NO₂ MW: 115.13 g/mol |
MDL Number:
MFCD09752973 CAS Number: 476014-92-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...