(3-Chloronaphthalen-2-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A1059099-100MG A1059099-1G A1059099-250MG
77782-999EA 125 USD
77782-999 77783-000 77783-001
(3-Chloronaphthalen-2-yl)boronic acid ≥95%
(3-Chloronaphthalen-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₈BClO₂
MW: 206.43 g/mol
MDL Number: MFCD18411294
CAS Number: 1873273-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR