tert-Butyl 3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)azetidine-1-carboxylate ≥95%

Supplier: Ambeed
Warning

A1058447-100MG A1058447-1G A1058447-250MG A1058447-50MG
77592-124EA 252.15 USD
77592-124 77592-126 77592-128 77619-758
tert-Butyl 3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)azetidine-1-carboxylate ≥95%
tert-Butyl 3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)azetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₂₀H₃₀BNO₅
MW: 375.27 g/mol
MDL Number: MFCD22683476
CAS Number: 1505515-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR