5-Methoxypyridine-2,3-diamine ≥97%, Ambeed.Inc

Supplier: Ambeed
Warning

A105683-1G A105683-100MG
77610-830EA 421.27 USD
77610-830 77682-971
5-Methoxypyridine-2,3-diamine ≥97%, Ambeed.Inc
5-Methoxypyridine-2,3-diamine

Precaución: For research use only.

Formula: C₆H₉N₃O
MW: 139.15 g/mol
CAS Number: 618439-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR