(S,E)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-phenylpent-4-enoic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-3-Styryl-Ala-OH

A105476-1G A105476-250MG
77684-954EA 125 USD
77684-954 77684-955
(S,E)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-phenylpent-4-enoic acid ≥98%
(S,E)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-phenylpent-4-enoic acid

Precaución: for Research Use Only

Formula: C₂₆H₂₃NO₄
MW: 413.46 g/mol
MDL Number: MFCD01311748
CAS Number: 159610-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR