Naphthol AS-D ≥98% (by HPLC)

Supplier: Ambeed
Danger

A105280-1000G A105280-500G A105280-100G
77599-538EA 41 USD
77599-538 77599-540 77666-540
Naphthol AS-D ≥98% (by HPLC)
Naphthol AS-D

Precaución: for Research Use Only

Formula: C₁₈H₁₅NO₂
MDL Number: MFCD00021634
CAS Number: 135-61-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR