Ethyl 5-ethyl-1H-imidazole-2-carboxylate ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A104858-1G A104858-250MG
77682-965EA 630 USD
77682-965 77682-966
Ethyl 5-ethyl-1H-imidazole-2-carboxylate ≥95%, Ambeed.Inc
Ethyl 5-ethyl-1H-imidazole-2-carboxylate

Precaución: For research use only.

Formula: C₈H₁₂N₂O₂
MW: 168.19 g/mol
MDL Number: MFCD24386911
CAS Number: 1171124-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR