Dimethyldipalmitylammonium bromide ≥98%

Supplier: Ambeed
Warning

A104718-100G A104718-25G
77623-872EA 194.67 USD
77623-872 77623-874
Dimethyldipalmitylammonium bromide ≥98%
Dimethyldipalmitylammonium bromide

Precaución: for Research Use Only

Formula: C₃₄H₇₂BrN
MDL Number: MFCD00191428
CAS Number: 70755-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR