3,5-Dimethylbenzene-1,2-diol ≥95%

Supplier: Ambeed
Warning

A1047123-100MG A1047123-1G
77782-981EA 144 USD
77782-981 77782-982
3,5-Dimethylbenzene-1,2-diol ≥95%
3,5-Dimethylbenzene-1,2-diol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MDL Number: MFCD24713456
CAS Number: 2785-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR