Ethylenediaminetetraacetic acid disodium zinc salt Zn 14.5-15.5%

Supplier: Ambeed
Warning

Synonyms: Disodium zinc ethylenediaminetetraacetate, Ethylenediaminetetraacetic Acid Disodium Zinc Salt, Ethylenediaminetetraacetic acid zinc disodium salt

A104698-500G A104698-1000G A104698-100G
77599-530EA 12.3 USD
77599-530 77666-522 77878-807
Ethylenediaminetetraacetic acid disodium zinc salt Zn 14.5-15.5%
Ethylenediaminetetraacetic acid disodium zinc salt

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂Na₂O₈Zn
MW: 399.60 g/mol
MDL Number: MFCD00135187
CAS Number: 14025-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR