2-(1-Ethyl-1H-pyrazol-4-yl)morpholine ≥95%

Supplier: Ambeed
Danger

A1046620-1G A1046620-50MG
77619-744EA 530.45 USD
77619-744 77782-976
2-(1-Ethyl-1H-pyrazol-4-yl)morpholine ≥95%
2-(1-Ethyl-1H-pyrazol-4-yl)morpholine

Precaución: for Research Use Only

Formula: C₉H₁₅N₃O
MDL Number: MFCD28023702
CAS Number: 1803600-81-8
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR