Dimethyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate ≥97%

Supplier: Ambeed
Warning

A104614-5G A104614-1G A104614-25G
77599-528EA 42.03 USD
77599-528 77666-518 77666-520
Dimethyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate ≥97%
Dimethyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₂O₆
MDL Number: MFCD00195480
CAS Number: 21881-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR