(S)-2-Phenylpyrrolidine hydrochloride ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A104596-250MG A104596-100MG A104596-1G
77682-963EA 95 USD
77682-963 77889-729 77889-730
(S)-2-Phenylpyrrolidine hydrochloride ≥95%, Ambeed.Inc
(S)-2-Phenylpyrrolidine hydrochloride

Precaución: For research use only.

Formula: C₁₀H₁₄ClN
MDL Number: MFCD12910597
CAS Number: 56523-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR