(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-hydroxybutanoic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Hse-OH

A1045738-1G A1045738-5G
77619-738EA 23.69 USD
77619-738 77619-740
(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-hydroxybutanoic acid ≥97%
(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-hydroxybutanoic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₉NO₅
MW: 341.3579 g/mol
MDL Number: MFCD02682651
CAS Number: 172525-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR