(7-Bromo-2,3-dihydrobenzofuran-2-yl)methanol ≥97%

Supplier: Ambeed
Warning

A104317-1G A104317-100MG
77592-104EA 179.38 USD
77592-104 77682-956
(7-Bromo-2,3-dihydrobenzofuran-2-yl)methanol ≥97%
(7-Bromo-2,3-dihydrobenzofuran-2-yl)methanol

Precaución: For research use only.

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
MDL Number: MFCD14705074
CAS Number: 852110-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR