Methyl 5-methylbenzo[d]oxazole-2-carboxylate ≥95%

Supplier: Ambeed
Warning

A104085-100MG A104085-250MG
77889-726EA 30 USD
77889-726 77889-727
Methyl 5-methylbenzo[d]oxazole-2-carboxylate ≥95%
Methyl 5-methylbenzo[d]oxazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
MDL Number: MFCD18909967
CAS Number: 27383-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR