(3S,4R)-rel-1-Boc-3-fluoro-4-(hydroxymethyl)piperidine ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3S,4R)-rel-tert-Butyl 3-fluoro-4-(hydroxymethyl)piperidine-1-carboxylate
A104034-100MG
A104034-250MG
A104034-1G
77592-098EA
90.2
USD
77592-098
77684-945
77891-767
(3S,4R)-rel-1-Boc-3-fluoro-4-(hydroxymethyl)piperidine ≥98%
(3S,4R)-rel-1-Boc-3-fluoro-4-(hydroxymethyl)piperidine
Precaución: for Research Use Only
Formula:
C₁₁H₂₀FNO₃ |
MDL Number:
MFCD24850020 CAS Number: 882033-93-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...