4-Bromo-2-nitro-1H-pyrrole ≥95%

Supplier: Ambeed
Warning

A1039526-250MG A1039526-1G
77570-230EA 293.55 USD
77570-230 77619-724
4-Bromo-2-nitro-1H-pyrrole ≥95%
4-Bromo-2-nitro-1H-pyrrole

Precaución: for Research Use Only

Formula: C₄H₃BrN₂O₂
MW: 190.98 g/mol
MDL Number: MFCD28661286
CAS Number: 30078-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR