4-Formyl-1-methylpyridin-1-ium iodide ≥95%

Supplier: Ambeed
Warning

A1039410-250MG A1039410-1G A1039410-5G
77619-720EA 61.8 USD
77619-720 77782-952 77782-953
4-Formyl-1-methylpyridin-1-ium iodide ≥95%
4-Formyl-1-methylpyridin-1-ium iodide

Precaución: for Research Use Only

Formula: C₇H₈INO
MDL Number: MFCD00955994
CAS Number: 13441-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR