(S)-2-(2-Methoxyphenyl)pyrrolidine hydrochloride ≥97%
Supplier: Ambeed
|
Warning
|
A103928-100MG
A103928-1G
A103928-250MG
77570-226EA
203
USD
77570-226
77747-777
77747-778
(S)-2-(2-Methoxyphenyl)pyrrolidine hydrochloride ≥97%
(S)-2-(2-Methoxyphenyl)pyrrolidine hydrochloride
Precaución: for Research Use Only
Formula:
C₁₁H₁₆ClNO |
MDL Number:
MFCD08751474 CAS Number: 1381928-83-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...