2-(5-Chloropyridin-2-yl)acetic acid ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A103904-100MG A103904-1G A103904-250MG A103904-5G
77682-951EA 22 USD
77682-951 77682-952 77884-946 77884-947
2-(5-Chloropyridin-2-yl)acetic acid ≥95%, Ambeed.Inc
2-(5-Chloropyridin-2-yl)acetic acid

Precaución: For research use only.

Formula: C₇H₆ClNO₂
MDL Number: MFCD09925072
CAS Number: 1000522-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR