2-(Prop-2-yn-1-yloxy)ethan-1-amine hydrochloride ≥95%
Supplier: Ambeed
|
Warning
|
A1038152-1G
A1038152-5G
77619-712EA
130.81
USD
77619-712
77619-714
2-(Prop-2-yn-1-yloxy)ethan-1-amine hydrochloride ≥95%
2-(Prop-2-yn-1-yloxy)ethan-1-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₀ClNO MW: 135.5900 g/mol |
MDL Number:
MFCD25970416 CAS Number: 1383604-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...