4-Bromo-1-cyclohexyl-1H-pyrazole ≥95%

Supplier: Ambeed
Warning

A1037608-1G A1037608-250MG
77619-710EA 292.52 USD
77619-710 77782-938
4-Bromo-1-cyclohexyl-1H-pyrazole ≥95%
4-Bromo-1-cyclohexyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₁₃BrN₂
MDL Number: MFCD12824339
CAS Number: 141302-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR