(E)-(4-Bromostyryl)boronic acid ≥97%

Supplier: Ambeed
Warning

A103564-100MG A103564-1G A103564-250MG A103564-50MG A103564-5G
77614-776EA 164.8 USD
77614-776 77747-771 77747-772 77747-773 77747-774
(E)-(4-Bromostyryl)boronic acid ≥97%
(E)-(4-Bromostyryl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₈BBrO₂
MW: 226.86 g/mol
CAS Number: 1795770-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR