(1-Cyanocyclopropyl)methanesulfonyl chloride ≥95%

Supplier: Ambeed
Danger

A1035347-1G A1035347-100MG A1035347-250MG
77570-208EA 842 USD
77570-208 77619-700 77782-928
(1-Cyanocyclopropyl)methanesulfonyl chloride ≥95%
(1-Cyanocyclopropyl)methanesulfonyl chloride

Precaución: for Research Use Only

Formula: C₅H₆ClNO₂S
MDL Number: MFCD25371603
CAS Number: 1461707-09-4
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR