4-((2,4-Dioxothiazolidin-5-ylidene)methyl)benzoic acid ≥95%

Supplier: Ambeed
Warning

A1034383-1G A1034383-250MG
77619-696EA 144.2 USD
77619-696 77619-698
4-((2,4-Dioxothiazolidin-5-ylidene)methyl)benzoic acid ≥95%
4-((2,4-Dioxothiazolidin-5-ylidene)methyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₇NO₄S
MW: 249.2426 g/mol
MDL Number: MFCD00297792
CAS Number: 199167-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR