4'-Chlorobiphenyl-4-carboxylic acid ≥95%

Supplier: Ambeed
Danger

Synonyms: 4'-Chloro[1,1'-biphenyl]-4-carboxylic acid, 4'-Chloro-[1,1'-biphenyl]-4-carboxylic acid

A103392-1G A103392-5G
77570-200EA 29.87 USD
77570-200 77684-940
4'-Chlorobiphenyl-4-carboxylic acid ≥95%
4'-Chlorobiphenyl-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₉ClO₂
MW: 232.67 g/mol
MDL Number: MFCD03424592
CAS Number: 5748-41-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR