(7-Chloroquinolin-8-yl)methanol ≥95%

Supplier: Ambeed
Warning

A1032340-100MG A1032340-250MG
77782-924EA 19 USD
77782-924 77782-925
(7-Chloroquinolin-8-yl)methanol ≥95%
(7-Chloroquinolin-8-yl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₈ClNO
MDL Number: MFCD24842811
CAS Number: 84163-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR