Quinizarin green ≥95%

Supplier: Ambeed
Warning

Synonyms: 1,4-Bis(p-tolylamino)anthracene-9,10-dione, Solvent Green 3, 1,4-Bis(p-toluidino)-9,10-anthraquinone, D and C Green No. 6

A103026-1000G A103026-25G A103026-500G
77666-486EA 306 USD
77666-486 77666-488 77666-490
Quinizarin green ≥95%
1,4-Bis(p-toluidino)-9,10-anthraquinone

Precaución: for Research Use Only

Formula: C₂₈H₂₂N₂O₂
MW: 418.496 g/mol
MDL Number: MFCD00001195
CAS Number: 128-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR