2,2-Dimethylpent-4-ynoic acid ≥95%

Supplier: Ambeed

A1029413-100MG A1029413-1G
77782-910EA 81 USD
77782-910 77782-911
2,2-Dimethylpent-4-ynoic acid ≥95%
2,2-Dimethylpent-4-ynoic acid

Precaución: for Research Use Only

Formula: C₇H₁₀O₂
MDL Number: MFCD19228208
CAS Number: 86101-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR