(S)-3-((tert-Butoxycarbonyl)amino)-6-methoxy-6-oxohexanoic acid ≥95%

Supplier: Ambeed
Warning

A1025864-1G A1025864-100MG A1025864-250MG
77570-146EA 44 USD
77570-146 77782-882 77782-883
(S)-3-((tert-Butoxycarbonyl)amino)-6-methoxy-6-oxohexanoic acid ≥95%
(S)-3-((tert-Butoxycarbonyl)amino)-6-methoxy-6-oxohexanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₆
MDL Number: MFCD29076497
CAS Number: 222846-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR