Azepane-1-carbonyl chloride ≥95%

Supplier: Ambeed
Danger

A1025068-5G A1025068-25G
77570-134EA 45.32 USD
77570-134 77619-682
Azepane-1-carbonyl chloride ≥95%
Azepane-1-carbonyl chloride

Precaución: for Research Use Only

Formula: C₇H₁₂ClNO
MW: 161.6293 g/mol
CAS Number: 27817-35-2
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR