5-(Difluoromethoxy)-2-methoxyaniline ≥95%

Supplier: Ambeed
Warning

A1020637-100MG A1020637-1G A1020637-250MG
77782-875EA 21 USD
77782-875 77782-876 77782-877
5-(Difluoromethoxy)-2-methoxyaniline ≥95%
5-(Difluoromethoxy)-2-methoxyaniline

Precaución: for Research Use Only

Formula: C₈H₉F₂NO₂
MDL Number: MFCD18389540
CAS Number: 1256627-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR