Dibenzyl 2-aminosuccinate 4-methylbenzenesulfonate ≥98%

Supplier: Ambeed
Warning

Synonyms: Dibenzyl aspartate 4-methylbenzenesulphonate

A101713-25G A101713-5G
77684-928EA 22 USD
77684-928 77684-929
Dibenzyl 2-aminosuccinate 4-methylbenzenesulfonate ≥98%
Dibenzyl 2-aminosuccinate 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₂₅H₂₇NO₇S
MW: 485.54 g/mol
MDL Number: MFCD00187195
CAS Number: 4079-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR