3-Fluoro-4-(trifluoromethyl)benzyl chloride ≥95%

Supplier: Ambeed
Danger

A1015192-1G A1015192-250MG A1015192-5G
77619-672EA 41.2 USD
77619-672 77619-674 77782-865
3-Fluoro-4-(trifluoromethyl)benzyl chloride ≥95%
3-Fluoro-4-(trifluoromethyl)benzyl chloride

Precaución: for Research Use Only

Formula: C₈H₅ClF₄
MDL Number: MFCD13185659
CAS Number: 444915-54-2
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR