2-Isocyanato-1,1-dimethoxyethane ≥95%

Supplier: Ambeed
Danger

A1013406-1G A1013406-250MG
77570-094EA 680 USD
77570-094 77570-096
2-Isocyanato-1,1-dimethoxyethane ≥95%
2-Isocyanato-1,1-dimethoxyethane

Precaución: for Research Use Only

Formula: C₅H₉NO₃
MDL Number: MFCD05864411
CAS Number: 71189-22-5
UN: 2206
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR