2-(trans-4-Aminocyclohexyl)acetonitrile hydrochloride ≥97%

Supplier: Ambeed
Warning

Synonyms: 2-[trans-4-aminocyclohexyl]acetonitrile hydrochloride

A101308-1G A101308-100MG
77614-744EA 219.39 USD
77614-744 77747-757
2-(trans-4-Aminocyclohexyl)acetonitrile hydrochloride ≥97%
2-(trans-4-Aminocyclohexyl)acetonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₅ClN₂
MW: 174.67 g/mol
MDL Number: MFCD30146461
CAS Number: 1313279-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR