Ruthenium(III) acetate ≥97%

Supplier: Ambeed
Danger

A1004809-100MG A1004809-1G A1004809-250MG
77619-536EA 17.51 USD
77619-536 77782-749 77782-750
Ruthenium(III) acetate ≥97%
Ruthenium(III) acetate

Precaución: for Research Use Only

Formula: C₆H₉O₆Ru
MDL Number: MFCD13194948
CAS Number: 72196-32-8
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR