1-Propylboronic acid pinacol ester ≥98%

Supplier: Ambeed
Danger

Synonyms: 4,4,5,5-Tetramethyl-2-propyl-1,3,2-dioxaborolane

A1004784-1G A1004784-25G A1004784-5G
77570-040EA 10 USD
77570-040 77782-744 77782-745
1-Propylboronic acid pinacol ester ≥98%
1-Propylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₉H₁₉BO₂
MDL Number: MFCD12546187
CAS Number: 67562-19-0
UN: 3272
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR