4-(trans-3-Amino-2,2,4,4-tetramethylcyclobutoxy)-2-chlorobenzonitrile hydrochloride ≥97%

Supplier: Ambeed
Warning

A1004519-250MG A1004519-5G
77595-654EA 91.23 USD
77595-654 77782-734
4-(trans-3-Amino-2,2,4,4-tetramethylcyclobutoxy)-2-chlorobenzonitrile hydrochloride ≥97%
4-(trans-3-Amino-2,2,4,4-tetramethylcyclobutoxy)-2-chlorobenzonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₂₀Cl₂N₂O
MDL Number: MFCD30729667
CAS Number: 1818885-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR