N,N-Dimethyldecan-1-amine oxide ≥98%

Supplier: Ambeed
Danger

A1004009-250MG A1004009-1G A1004009-5G
77619-518EA 21.63 USD
77619-518 77782-730 77782-731
N,N-Dimethyldecan-1-amine oxide ≥98%
N,N-Dimethyldecan-1-amine oxide

Precaución: for Research Use Only

Formula: C₁₂H₂₇NO
MDL Number: MFCD00082523
CAS Number: 2605-79-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR