Tetrakis(4-N,N-dimethylaminophenyl)porphyrin ≥97%

Supplier: Ambeed
Warning

A1003744-100MG A1003744-1G
77570-032EA 55.62 USD
77570-032 77619-516
Tetrakis(4-N,N-dimethylaminophenyl)porphyrin ≥97%
Tetrakis(4-N,N-dimethylaminophenyl)porphyrin

Precaución: for Research Use Only

Formula: C₅₂H₅₀N₈
MW: 787.0070 g/mol
MDL Number: MFCD01074432
CAS Number: 14945-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR